missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Dibromo-2-hydroxynaphthalene-3-carboxylic acid, 97%

$61.72 - $233.97

Chemical Identifiers

CAS 1779-10-8
Molecular Formula C11H6Br2O3
Formula Weight 345.98
MDL Number MFCD00046367
Beilstein 2378969
Synonym 1,6-Dibromo-2-naphthol-3-carboxylic acid
Chemical Name or Material 1,6-Dibromo-2-hydroxynaphthalene-3-carboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0701914 SDS Alfa Aesar™
L0701914
25g Each for $233.97
AAL0701906 SDS Alfa Aesar™
L0701906
5g Each for $61.72
Specifications Description & Specifications

Chemical Identifiers

1779-10-8
345.98
2378969
1,6-Dibromo-2-hydroxynaphthalene-3-carboxylic acid
C11H6Br2O3
MFCD00046367
1,6-Dibromo-2-naphthol-3-carboxylic acid

Specifications

1779-10-8
25g
Odorless
MFCD00046367
1,6-Dibromo-2-naphthol-3-carboxylic acid
97%
249° to 253°C
C11H6Br2O3
345.98
2378969
1,6-Dibromo-2-hydroxynaphthalene-3-carboxylic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

TSCA : TSCA