Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Diaminohexane, 98+%

$26.68 - $73.08

Chemical Identifiers

Chemical Name or Material 1,6-Diaminohexane
CAS 124-09-4
Molecular Formula C6H16N2
Linear Formula H2N(CH2)6NH2
Formula Weight 116.21
MDL Number MFCD00008243
UN Number UN2280
Beilstein 1098307
Merck Index 14,4695
Synonym Hexamethylenediamine; 1,6-Hexanediamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA142120E

 
alfa aesar™
A142120E
2500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1421236

 
alfa aesar™
A1421236
500g Each for $73.08

AAA1421222

 
alfa aesar™
A1421222
100g Each for $26.68
Description & Specifications

Specifications

Formula Weight 116.21
MDL Number MFCD00008243
Flash Point 85°C (185°F)
Quantity 2500g
Linear Formula H2N(CH2)6NH2
UN Number UN2280
Merck Index 14,4695
RTECS Number MO1180000
Assay ≥98%
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: HEXAMETHYLENEDIAMINE, SOLID
Molecular Formula C6H16N2
Beilstein 1098307
Health Hazard 1 Danger
Chemical Name or Material 1,6-Diaminohexane
Boiling Point 199° to 204°C
Melting Point 39° to 43°C
Synonym Hexamethylenediamine; 1,6-Hexanediamine
Odor Amine-like
pH 12.4
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
TSCA TSCA
CAS 124-09-4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P309-P310
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Applications
1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.

Solubility
Soluble in water. Slightly soluble in ether, alcohol and benzene.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, strong oxidizing agents, acid chlorides, carbon dioxide and acid anhydrides.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P309-P310
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.