missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Diacetoxyhexane, 98%

$93.80 - $312.65

Chemical Identifiers

CAS 6222-17-9
Molecular Formula CH3CO2(CH2)6O2CCH3
Formula Weight 202.25
MDL Number MFCD00191655
Synonym 1,6-Hexanediol diacetate
Chemical Name or Material 1,6-Diacetoxyhexane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2153118 SDS Alfa Aesar™
B2153118
50g Each for $93.80
AAB2153130 SDS Alfa Aesar™
B2153130
250g Each for $312.65
Specifications Description & Specifications

Chemical Identifiers

6222-17-9
202.25
1,6-Hexanediol diacetate
CH3CO2(CH2)6O2CCH3
MFCD00191655
1,6-Diacetoxyhexane

Specifications

50g
CH3CO2(CH2)6O2CCH3
MFCD00191655
103° to 105°C (6mmHg)
98%
6222-17-9
202.25
1,6-Hexanediol diacetate
1,6-Diacetoxyhexane

Safety and Handling

TSCA : TSCA