Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(6-Chloro-2-pyridyl)-1H-3-hydroxy-1,2,4-triazole

$301.04 - $301.04

Chemical Identifiers

Chemical Name or Material 1-(6-Chloro-2-pyridyl)-1H-3-hydroxy-1,2,4-triazole
CAS 80714-26-7
Molecular Formula C7H5ClN4O
Formula Weight 196.60g/mol
MDL Number MFCD00138720
Synonym 1-(6-Chloro-2-pyridyl)-1,2,4-triazol-3-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5019503

 
alfa aesar™
H50195-03
Each for $301.04
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 1-(6-Chloro-2-pyridyl)-1H-3-hydroxy-1,2,4-triazole
CAS 80714-26-7
Molecular Formula C7H5ClN4O
Formula Weight 196.60g/mol
MDL Number MFCD00138720
Synonym 1-(6-Chloro-2-pyridyl)-1,2,4-triazol-3-ol