missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,6-Bis(diphenylphosphino)hexane, 97%

Manufacturer:  Alfa Aesar™ H5552806

 View more versions of this product

Catalog No. AAH5552806



Specifications

Specifications

1,6-Bis(diphenylphosphino)hexane
5g
C30H32P2
MFCD00003053
GPORFKPYXATYNX-UHFFFAOYSA-N
6-diphenylphosphanylhexyl(diphenyl)phosphane
2754312
97%
124°C to 126°C
19845-69-3
(C6H5)2P(CH2)6P(C6H5)2
1,6-bis diphenylphosino hexane
C1=CC=C(C=C1)P(CCCCCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
454.534
454.53

Chemical Identifiers

19845-69-3
454.534
GPORFKPYXATYNX-UHFFFAOYSA-N
2754312
C1=CC=C(C=C1)P(CCCCCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
C30H32P2
MFCD00003053
1,6-bis diphenylphosino hexane
6-diphenylphosphanylhexyl(diphenyl)phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents