missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane, TCI America™

$73.33 - $221.53

Chemical Identifiers

CAS 2264-01-9
Molecular Formula C12H10F8O4
Molecular Weight (g/mol) 370.20
MDL Number MFCD00078412
InChI Key BWTMTZBMAGYMOD-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-Octafluorohexamethylene Diacrylate, 2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol Diacrylate
PubChem CID 2775863
IUPAC Name (2,2,3,3,4,4,5,5-octafluoro-6-prop-2-enoyloxyhexyl) prop-2-enoate
SMILES C=CC(=O)OCC(C(C(C(COC(=O)C=C)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B23401G View Documents TCI America
B23401G
1g Each for $73.33
 
B23405G View Documents TCI America
B23405G
5g Each for $221.53
 
Specifications

Chemical Identifiers

2264-01-9
370.20
BWTMTZBMAGYMOD-UHFFFAOYSA-N
2775863
C=CC(=O)OCC(C(C(C(COC(=O)C=C)(F)F)(F)F)(F)F)(F)F
C12H10F8O4
MFCD00078412
2,2,3,3,4,4,5,5-Octafluorohexamethylene Diacrylate, 2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol Diacrylate
(2,2,3,3,4,4,5,5-octafluoro-6-prop-2-enoyloxyhexyl) prop-2-enoate

Specifications

Yellow
2264-01-9
MFCD00078412
2,2,3,3,4,4,5,5-Octafluorohexamethylene Diacrylate, 2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol Diacrylate
C=CC(=O)OCC(C(C(C(COC(=O)C=C)(F)F)(F)F)(F)F)(F)F
370.20
370.20
≥90.0% (GC)
1g
C12H10F8O4
3532
BWTMTZBMAGYMOD-UHFFFAOYSA-N
(2,2,3,3,4,4,5,5-octafluoro-6-prop-2-enoyloxyhexyl) prop-2-enoate
2775863
Clear Liquid at 20°C
1,6-Bis(acryloyloxy)-2,2,3,3,4,4,5,5-octafluorohexane
Safety and Handling

Safety and Handling

TSCA : No