missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,6-Anhydro-β-D-glucose-2,3,4-tri-O-acetate, 98+%, ACROS Organics™
Chemical Identifiers
CAS | 13242-55-2 |
---|---|
Molecular Formula | C12H16O8 |
Molecular Weight (g/mol) | 288.26 |
MDL Number | MFCD00063247 |
InChI Key | BAKQMOSGYGQJOJ-UHFZAUJKSA-N |
Synonym | 1,6-anhydro-, 2,3,4-tri-o-acetyl-1,6-anhydro-, 2r,3s,4r,5r-2,3-bis acetyloxy-6,8-dioxabicyclo 3.2.1 octan-4-yl acetate, a-d-glucopyranose, a-d-glucopyranose 2,3,4-triacetate, triacetyllevoglucosan |
PubChem CID | 134159065 |
IUPAC Name | [(1S,2S,3S,4S,5R)-3,4-diacetyloxy-6,8-dioxabicyclo[3.2.1]octan-2-yl] acetate |
SMILES | CC(=O)OC1C2COC(O2)C(C1OC(=O)C)OC(=O)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC353685000
|
Thermo Fisher Scientific
353685000 |
500mg | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
13242-55-2 | |
288.26 | |
BAKQMOSGYGQJOJ-UHFZAUJKSA-N | |
134159065 | |
CC(=O)OC1C2COC(O2)C(C1OC(=O)C)OC(=O)C |
C12H16O8 | |
MFCD00063247 | |
1,6-anhydro-, 2,3,4-tri-o-acetyl-1,6-anhydro-, 2r,3s,4r,5r-2,3-bis acetyloxy-6,8-dioxabicyclo 3.2.1 octan-4-yl acetate, a-d-glucopyranose, a-d-glucopyranose 2,3,4-triacetate, triacetyllevoglucosan | |
[(1S,2S,3S,4S,5R)-3,4-diacetyloxy-6,8-dioxabicyclo[3.2.1]octan-2-yl] acetate |
Specifications
Authentic | |
− 66.00 | |
111.0°C to 112.0°C | |
13242-55-2 | |
100.0 | |
C12H16O8 | |
1,6-anhydro-, 2,3,4-tri-o-acetyl-1,6-anhydro-, 2r,3s,4r,5r-2,3-bis acetyloxy-6,8-dioxabicyclo 3.2.1 octan-4-yl acetate, a-d-glucopyranose, a-d-glucopyranose 2,3,4-triacetate, triacetyllevoglucosan | |
BAKQMOSGYGQJOJ-UHFZAUJKSA-N | |
[(1S,2S,3S,4S,5R)-3,4-diacetyloxy-6,8-dioxabicyclo[3.2.1]octan-2-yl] acetate | |
134159065 | |
Crystalline Powder | |
1,6-Anhydro-β-D-glucose-2,3,4-tri-O-acetate, 99% |
Glass bottle | |
White | |
500mg | |
98.0 | |
98+% | |
MFCD00063247 | |
Solubility in water: soluble. | |
CC(=O)OC1C2COC(O2)C(C1OC(=O)C)OC(=O)C | |
288.26 | |
288.26 | |
98+% |
Safety and Handling
EINECSNumber : 236-222-1
TSCA : TSCA