missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-[5-(Trifluoromethyl)-2-pyridyl]-1,4-diazepane,95+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals AW00291DA
Specifications
1-[5-(Trifluoromethyl)-2-pyridyl]-1, 4-diazepane | |
97% | |
C11H14F3N3 | |
1g | |
IBMSHQVIAUTKDL-UHFFFAOYSA-N | |
1-[5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane | |
2760779 | |
97% |
306934-70-3 | |
Amber Glass Bottle | |
MFCD00661783 | |
1-5-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazepane, 1-5-trifluoromethyl pyrid-2-yl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4 diazepane, 1h-1,4-diazepine,hexahydro-1-5-trifluoromethyl-2-pyridinyl, 1-5-trifluoromethyl-pyridin-2-yl 1,4-diazepane, 1-5-trifluoromethyl-2-pyridinyl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazaperhydroepine | |
FC(F)(F)C1=CN=C(C=C1)N1CCCNCC1 | |
245.25 | |
245.25 |
Chemical Identifiers
306934-70-3 | |
245.25 | |
IBMSHQVIAUTKDL-UHFFFAOYSA-N | |
2760779 | |
FC(F)(F)C1=CN=C(C=C1)N1CCCNCC1 |
C11H14F3N3 | |
MFCD00661783 | |
1-5-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazepane, 1-5-trifluoromethyl pyrid-2-yl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4 diazepane, 1h-1,4-diazepine,hexahydro-1-5-trifluoromethyl-2-pyridinyl, 1-5-trifluoromethyl-pyridin-2-yl 1,4-diazepane, 1-5-trifluoromethyl-2-pyridinyl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazaperhydroepine | |
1-[5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane |
Safety and Handling
GHS Signal Word: Danger