Please login to your online account to display your discounted pricing

1-[5-(Trifluoromethyl)-2-pyridyl]-1,4-diazepane,95+% Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  AW00291DE

 View more versions of this product

Catalog No. AW00291DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 245.25
Quantity 5g
Assay 97%
Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridyl]-1, 4-diazepane
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C11H14F3N3
CAS 306934-70-3
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridyl]-1, 4-diazepane
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H14F3N3
Formula Weight 245.25
Accessories

GHS Signal Word: Danger