Please login to your online account to display your discounted pricing

1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC367100050

 View more versions of this product

Catalog No. AC367100050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Chunks
Formula Weight 231.22
Quantity 5g
Color Slightly yellow
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine
CAS 132834-58-3
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
CAS Min % 97.5
Molecular Formula C10H12F3N3
MDL Number MFCD00114708
Assay Percent Range 98%

Specifications

Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine
Name Note 98%
CAS 132834-58-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12F3N3
Formula Weight 231.22
MDL Number MFCD00114708
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents