Please login to your online account to display your discounted pricing

1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC367100010

 View more versions of this product

Catalog No. AC367100010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Chunks
Formula Weight 231.22
Quantity 1g
Color Slightly yellow
CAS Max % 100.0
Packaging Glass bottle
CAS 132834-58-3
Molecular Formula C10H12F3N3
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
MDL Number MFCD00114708
CAS Min % 97.5
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Name Note 98%
Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine
Name Note 98%
CAS 132834-58-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12F3N3
Formula Weight 231.22
MDL Number MFCD00114708
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents