missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine, 98%, ACROS Organics™

Chemical Identifiers

CAS 132834-58-3
Molecular Formula C10H14F3N3
Molecular Weight (g/mol) 233.24
MDL Number MFCD00114708
InChI Key BNMSJUIMZULLAS-UHFFFAOYSA-P
Synonym 1-5-trifluoromethyl pyrid-2-yl piperazine, 1-5-trifluoromethyl pyridin-2-yl piperazine, 1-5-trifluoromethyl pyridine-2-yl piperazine, 1-5-trifluoromethyl-2-pyridinyl piperazine, 1-5-trifluoromethyl-2-pyridinyl-piperazine, 1-5-trifluoromethyl-2-pyridyl piperazine, 1-5-trifluoromethyl-pyridin-2-yl-piperazine, 5-trifluoromethyl-2-pyridyl piperazine, piperazine, 1-5-trifluoromethyl-2-pyridinyl
PubChem CID 667618
IUPAC Name 1-[5-(trifluoromethyl)pyridin-2-yl]piperazine
SMILES FC(F)(F)C1=CC=C([NH+]=C1)N1CC[NH2+]CC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367100010
SDS View Documents
Thermo Fisher Scientific
AC367100010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

132834-58-3
233.24
BNMSJUIMZULLAS-UHFFFAOYSA-P
667618
FC(F)(F)C1=CC=C([NH+]=C1)N1CC[NH2+]CC1
C10H14F3N3
MFCD00114708
1-5-trifluoromethyl pyrid-2-yl piperazine, 1-5-trifluoromethyl pyridin-2-yl piperazine, 1-5-trifluoromethyl pyridine-2-yl piperazine, 1-5-trifluoromethyl-2-pyridinyl piperazine, 1-5-trifluoromethyl-2-pyridinyl-piperazine, 1-5-trifluoromethyl-2-pyridyl piperazine, 1-5-trifluoromethyl-pyridin-2-yl-piperazine, 5-trifluoromethyl-2-pyridyl piperazine, piperazine, 1-5-trifluoromethyl-2-pyridinyl
1-[5-(trifluoromethyl)pyridin-2-yl]piperazine

Specifications

Authentic
122.0°C (2.0 mmHg)
1g
97.5
97.5% min. (GC)
MFCD00114708
BNMSJUIMZULLAS-UHFFFAOYSA-P
1-[5-(trifluoromethyl)pyridin-2-yl]piperazine
667618
Crystalline Chunks
1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine, 98%
Glass bottle
Yellow
132834-58-3
100.0
C10H14F3N3
1-5-trifluoromethyl pyrid-2-yl piperazine, 1-5-trifluoromethyl pyridin-2-yl piperazine, 1-5-trifluoromethyl pyridine-2-yl piperazine, 1-5-trifluoromethyl-2-pyridinyl piperazine, 1-5-trifluoromethyl-2-pyridinyl-piperazine, 1-5-trifluoromethyl-2-pyridyl piperazine, 1-5-trifluoromethyl-pyridin-2-yl-piperazine, 5-trifluoromethyl-2-pyridyl piperazine, piperazine, 1-5-trifluoromethyl-2-pyridinyl
FC(F)(F)C1=CC=C([NH+]=C1)N1CC[NH2+]CC1
233.24
231.22
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Product Certifications