missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,5-Pentamethylene-1H-tetrazole, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1618514

 View more versions of this product

Catalog No. AAL1618514



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,5-Pentamethylene-1H-tetrazole
59°C to 61°C
C6H10N4
25 g
135492
14,7141
CWRVKFFCRWGWCS-UHFFFAOYSA-N
6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
5917
138.17
54-95-5
194°C (12 mmHg)
MFCD00005939
UN2811
Hygroscopic
pentylenetetrazol, pentylenetetrazole, pentetrazol, metrazole, cardiazol, cardiazole, cardiol, corazol, 1,5-pentamethylenetetrazole, corazole
C1CCC2=NN=NN2CC1
138.174
CHEBI:34910
98%

Chemical Identifiers

54-95-5
138.174
CWRVKFFCRWGWCS-UHFFFAOYSA-N
5917
6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
C6H10N4
MFCD00005939
pentylenetetrazol, pentylenetetrazole, pentetrazol, metrazole, cardiazol, cardiazole, cardiol, corazol, 1,5-pentamethylenetetrazole, corazole
CHEBI:34910
C1CCC2=NN=NN2CC1
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 200-219-3

RTECSNumber : XF8225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only