Learn More
1,5-Naphthyridine, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H64484MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,5-Naphthyridine | |
69°C to 75°C | |
1.25 g/mL | |
MFCD00039727 | |
109350 | |
VMLKTERJLVWEJJ-UHFFFAOYSA-N | |
1,5-naphthyridine | |
136070 | |
130.15 | |
Solid |
254-79-5 | |
White | |
C8H6N2 | |
250 mg | |
1,5-diazanaphthalene, 1,5-pyridopyridine, pyridino 3,2-b pyridine, pyridopyridine, 1,5-naphtyridine, 1,5 naphthyridine, zlchem 698, pubchem18072, acmc-209gka, pyrido 1,5-b pyridine | |
C1=CC2=NC=CC=C2N=C1 | |
130.15 | |
CHEBI:36625 | |
≥97% |
Chemical Identifiers
254-79-5 | |
130.15 | |
VMLKTERJLVWEJJ-UHFFFAOYSA-N | |
136070 | |
1,5-naphthyridine |
C8H6N2 | |
MFCD00039727 | |
1,5-diazanaphthalene, 1,5-pyridopyridine, pyridino 3,2-b pyridine, pyridopyridine, 1,5-naphtyridine, 1,5 naphthyridine, zlchem 698, pubchem18072, acmc-209gka, pyrido 1,5-b pyridine | |
CHEBI:36625 | |
C1=CC2=NC=CC=C2N=C1 |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only