Please login to your online account to display your discounted pricing

1,5-Naphthalenedisulfonic acid disodium salt, dihydrate, 98%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  415215000

 View more versions of this product

Catalog No. AC415215000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 368.29
Physical Form Crystalline Powder
Quantity 500g
Color White to light beige
TSCA TSCA
EINECS Number 216-732-0
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 97.5
CAS 36511-65-6
Chemical Name or Material 1, 5-Naphthalenedisulfonic acid disodium salt, dihydrate
Assay Percent Range 98%
Name Note 98%
Molecular Formula C10H6Na2O6S2·2H2O
CAS 76758-30-0

Specifications

Chemical Name or Material 1, 5-Naphthalenedisulfonic acid disodium salt, dihydrate
Name Note 98%
CAS 76758-30-0
CAS Min % 97.5
CAS Max % 100.0
CAS 36511-65-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H6Na2O6S2·2H2O
Formula Weight 368.29