Please login to your online account to display your discounted pricing

1,5-Naphthalenedisulfonic acid disodium salt, dihydrate, 98%, ACROS Organics™

$385.03 - $385.03

Chemical Identifiers

Chemical Name or Material 1, 5-Naphthalenedisulfonic acid disodium salt, dihydrate
Name Note 98%
CAS 76758-30-0
CAS Min % 97.5
CAS Max % 100.0
CAS 36511-65-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H6Na2O6S2·2H2O
Formula Weight 368.29
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC415215000

  SDS
thermo fisher scientific
415215000
500g Each for $385.03
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 368.29
Quantity 500g
Color White to light beige
EINECS Number 216-732-0
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material 1, 5-Naphthalenedisulfonic acid disodium salt, dihydrate
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H6Na2O6S2·2H2O
CAS 76758-30-0
CAS Min % 97.5
CAS 36511-65-6