Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(5-Methyl-2-pyridyl)piperazine, 97%

$98.39 - $342.04

Chemical Identifiers

Chemical Name or Material 1-(5-Methyl-2-pyridyl)piperazine
CAS 104395-86-0
Molecular Formula C10H15N3
Formula Weight 177.25
MDL Number MFCD08061050
Synonym 1-(5-Methylpyridin-2-yl)piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50897MD

  SDS
alfa aesar™
H50897MD
250mg Each for $98.39

AAH5089703

  SDS
alfa aesar™
H5089703
1g Each for $342.04
Description & Specifications

Specifications

Formula Weight 177.25
CAS 104395-86-0
Assay 97%
MDL Number MFCD08061050
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-(5-Methyl-2-pyridyl)piperazine
Molecular Formula C10H15N3
Synonym 1-(5-Methylpyridin-2-yl)piperazine
Health Hazard 1 Warning
Quantity 250mg

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.