Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(5-Iodo-2-pyridyl)piperazine, 95%

$77.15 - $171.77

Chemical Identifiers

Chemical Name or Material 1-(5-Iodo-2-pyridyl)piperazine
CAS 219635-89-9
Molecular Formula C9H12IN3
Formula Weight 289.12
MDL Number MFCD04039867
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2728903

  SDS
alfa aesar™
H2728903
1g Each for $171.77

AAH27289MD

  SDS
alfa aesar™
H27289MD
250mg Each for $77.15
Description & Specifications

Specifications

Formula Weight 289.12
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point ≈135°C
Quantity 1g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H12IN3
Health Hazard 1 Danger
CAS 219635-89-9
Assay 95%
Chemical Name or Material 1-(5-Iodo-2-pyridyl)piperazine
MDL Number MFCD04039867
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.