missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Hexadiyne, 50% in pentane

Manufacturer:  Alfa Aesar™ 1872009

 View more versions of this product

Catalog No. AA1872009



Description

Description

  • Packaged under argon
Specifications

Specifications

1,5-Hexadiyne
−2° (29°F)
628-16-0
MFCD00014924
1,5-hexadiyne, dipropargyl, bipropargyl, hc#cch2ch2c#ch, 1,5-hexadiyne in pentane, hccch2ch2cch, acmc-1b142, 1,5-hexadiyne 6ci, 8ci, 9ci, 1,5-hexadiyne solution, 50 wt. % in pentane 1g
YFIBSNDOVCWPBL-UHFFFAOYSA-N
hexa-1,5-diyne
69402
78.11
0.805g/mL
10g
C6H6
UN3295
50% in pentane
C#CCCC#C
78.114
CHEBI:37821
Liquid

Chemical Identifiers

628-16-0
78.114
YFIBSNDOVCWPBL-UHFFFAOYSA-N
69402
hexa-1,5-diyne
C6H6
MFCD00014924
1,5-hexadiyne, dipropargyl, bipropargyl, hc#cch2ch2c#ch, 1,5-hexadiyne in pentane, hccch2ch2cch, acmc-1b142, 1,5-hexadiyne 6ci, 8ci, 9ci, 1,5-hexadiyne solution, 50 wt. % in pentane 1g
CHEBI:37821
C#CCCC#C
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

SDS Documents