Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Hexadien-3-ol, 95%

$50.18 - $50.18

Chemical Identifiers

Chemical Name or Material 1,5-Hexadien-3-ol
CAS 924-41-4
Molecular Formula C6H10O
Formula Weight 98.15
MDL Number MFCD00008662
UN Number UN1987
Beilstein 1719962
Synonym Allyl vinyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0318204

  SDS
alfa aesar™
L0318204*
2g Each for $50.18
Description & Specifications

Specifications

Formula Weight 98.15
Chemical Name or Material 1,5-Hexadien-3-ol
Assay 95%
MDL Number MFCD00008662
CAS 924-41-4
UN Number UN1987
Beilstein 1719962
Synonym Allyl vinyl carbinol
Boiling Point 133° to 134°C
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Flash Point 29°C (84°F)
Health Hazard 3 GHS P Statement
P210-P261
Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Quantity 2g
Molecular Formula C6H10O
Health Hazard 1 Warning

Applications
1,5-Hexadien-3-ol is used in the synthesis of (+)-obtusenyne. It is used as intermediates. It undergoes Sharpless kinetic resolution and ring-closing metathesis to yield nine membered oxocene.

Solubility
Sparingly Soluble.

Notes
Keep away from heat. Incompatible with Oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261
Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS H Statement
H226
Flammable liquid and vapor.