missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Diphenylcarbazide, ACS

Manufacturer:  Alfa Aesar™ 3139014

 View more versions of this product

Catalog No. AA3139014



Specifications

Specifications

1,5-Diphenylcarbazide
White to Cream
25g
C13H14N4O
752039
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
KSPIHGBHKVISFI-UHFFFAOYSA-N
1,3-dianilinourea
8789
242.28
ACS
Air & Light Sensitive
170°C to 176°C
140-22-7
MFCD00003013
14,3321
Soluble in hot alcohol, acetone, glacial acetic acid
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
242.282
CHEBI:4641
Crystalline

Chemical Identifiers

140-22-7
242.282
KSPIHGBHKVISFI-UHFFFAOYSA-N
8789
1,3-dianilinourea
C13H14N4O
MFCD00003013
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
CHEBI:4641
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

RTECSNumber : FF2750000

TSCA : TSCA

SDS Documents