Please login to your online account to display your discounted pricing

1,5-Dimethyl-2-pyrrolecarbonitrile, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 5-Dimethyl-2-pyrrolecarbonitrile
Name Note 99%
CAS 56341-36-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8N2
Formula Weight 120.15
MDL Number MFCD00003086
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC189980500

  SDS
thermo fisher scientific
189980500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 120.15
Quantity 50g
Color Brown-gray
TSCA TSCA
EINECS Number 260-120-6
Melting Point 53° to 56°C
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Chemical Name or Material 1, 5-Dimethyl-2-pyrrolecarbonitrile
MDL Number MFCD00003086
Name Note 99%
CAS 56341-36-7
Assay Percent Range 99%
Molecular Formula C7H8N2
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.