Please login to your online account to display your discounted pricing

(1,5-Dimethyl-1H-pyrrol-2-yl)methylamine, 95+%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  SP01447CB

 View more versions of this product

Catalog No. SP01447CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 124.19
Quantity 250mg
Assay 90%
Chemical Name or Material (1, 5-Dimethyl-1H-pyrrol-2-yl)methylamine
Assay Percent Range 90%
Molecular Formula C7H12N2
CAS 118799-24-9

Specifications

Chemical Name or Material (1, 5-Dimethyl-1H-pyrrol-2-yl)methylamine
Assay 90%
Assay Percent Range 90%
Molecular Formula C7H12N2
Formula Weight 124.19