Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Dimethyl-1H-pyrazole-3-carboxylic acid, 98%

Chemical Identifiers

Chemical Name or Material 1,5-Dimethyl-1H-pyrazole-3-carboxylic acid
CAS 5744-59-2
Formula Weight 140.14
MDL Number MFCD00085040
Beilstein 115917
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6447503

 
alfa aesar™
H6447503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6447506

 
alfa aesar™
H6447506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6447514

 
alfa aesar™
H6447514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 170° to 174°C
Formula Weight 140.14
Quantity 1g
Physical Form Solid
MDL Number MFCD00085040
CAS 5744-59-2
Beilstein 115917
Storage Note 1 2° to 8°C
Chemical Name or Material 1,5-Dimethyl-1H-pyrazole-3-carboxylic acid
Assay 98%
Safety and Handling

2° to 8°C