missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,5-Dimethyl-1H-pyrazole-3-carboxylic acid, 95+%, Thermo Scientific™
Chemical Identifiers
CAS | 5744-59-2 |
---|---|
Molecular Formula | C6H8N2O2 |
Molecular Weight (g/mol) | 140.14 |
MDL Number | MFCD00085040 |
InChI Key | PXRXGHUTKHXUGF-UHFFFAOYSA-N |
Synonym | 1,5-dimethyl-1h-pyrazole-3-carboxylic acid, 1,5-dimethyl-1h-pyrazole-3-carboxylicacid, 1,5-dimethyl-3-pyrazolecarboxylic acid, 1h-pyrazole-3-carboxylic acid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic, acmc-209lyb, pyrazole-3-carboxylic acid, 1,5-dimethyl, 1h-pyrazole-3-carboxylicacid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid # |
PubChem CID | 587757 |
IUPAC Name | 1,5-dimethylpyrazole-3-carboxylic acid |
SMILES | CN1N=C(C=C1C)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
CC12601DA
|
Thermo Scientific Chemicals
CC12601DA |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC12601EA
|
Thermo Scientific Chemicals
CC12601EA |
10g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
5744-59-2 | |
140.14 | |
PXRXGHUTKHXUGF-UHFFFAOYSA-N | |
587757 | |
CN1N=C(C=C1C)C(O)=O |
C6H8N2O2 | |
MFCD00085040 | |
1,5-dimethyl-1h-pyrazole-3-carboxylic acid, 1,5-dimethyl-1h-pyrazole-3-carboxylicacid, 1,5-dimethyl-3-pyrazolecarboxylic acid, 1h-pyrazole-3-carboxylic acid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic, acmc-209lyb, pyrazole-3-carboxylic acid, 1,5-dimethyl, 1h-pyrazole-3-carboxylicacid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid # | |
1,5-dimethylpyrazole-3-carboxylic acid |
Specifications
5744-59-2 | |
C6H8N2O2 | |
1g | |
PXRXGHUTKHXUGF-UHFFFAOYSA-N | |
1,5-dimethylpyrazole-3-carboxylic acid | |
587757 | |
95% |
95% | |
MFCD00085040 | |
1,5-dimethyl-1h-pyrazole-3-carboxylic acid, 1,5-dimethyl-1h-pyrazole-3-carboxylicacid, 1,5-dimethyl-3-pyrazolecarboxylic acid, 1h-pyrazole-3-carboxylic acid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic, acmc-209lyb, pyrazole-3-carboxylic acid, 1,5-dimethyl, 1h-pyrazole-3-carboxylicacid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid # | |
CN1N=C(C=C1C)C(O)=O | |
140.14 | |
140.14 | |
1, 5-Dimethyl-1H-pyrazole-3-carboxylic acid |
Safety and Handling
Warning