Please login to your online account to display your discounted pricing

1,5-Dimethyl-1H-pyrazole-3-carboxylic acid, 95+%, Maybridge


Quantity 25g

Manufacturer: fisher scientific company  CC12601EB

 View more versions of this product

Catalog No. CC12601EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 140.14
Quantity 25g
Assay 95%
Molecular Formula C6H8N2O2
Chemical Name or Material 1, 5-Dimethyl-1H-pyrazole-3-carboxylic acid
Assay Percent Range 95%
CAS 5744-59-2
Health Hazard 1 Warning

Specifications

Chemical Name or Material 1, 5-Dimethyl-1H-pyrazole-3-carboxylic acid
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H8N2O2
Formula Weight 140.14
Accessories

Warning