Please login to your online account to display your discounted pricing

(1,5-Dimethyl-1H-pyrazol-3-yl)methylamine, 97%, Maybridge


Quantity 250mg
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC12613CB

 View more versions of this product

Catalog No. CC12613CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 125.17
Quantity 250mg
Assay 97%
CAS Max % 100.0
Packaging Amber glass bottle
Assay Percent Range 97%
Chemical Name or Material (1, 5-Dimethyl-1H-pyrazol-3-yl)methylamine
Molecular Formula C6H11N3
MDL Number MFCD03422531
CAS 423768-52-9
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (1, 5-Dimethyl-1H-pyrazol-3-yl)methylamine
CAS 423768-52-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H11N3
Formula Weight 125.17
MDL Number MFCD03422531
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.