missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1,5-Dimethyl-1H-pyrazol-3-yl)methylamine, 97%, Maybridge

Manufacturer:  Thermo Scientific CC12613DE

 View more versions of this product

Catalog No. CC12613DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(1, 5-Dimethyl-1H-pyrazol-3-yl)methylamine
423768-52-9
100.0
C6H11N3
1,5-dimethyl-1h-pyrazol-3-yl methanamine, 1,5-dimethyl-1h-pyrazol-3-yl methylamine, 3-aminomethyl-1,5-dimethylpyrazole, 1-1,5-dimethyl-1h-pyrazol-3-yl methanamine, 1,5-dimethylpyrazol-3-yl methanamine, 3-aminomethyl-1,5-dimethyl-1h-pyrazole, 1,5-dimethylpyrazol-3-yl methylamine, 1h-pyrazole-3-methanamine,1,5-dimethyl, 1-1,5-dimethylpyrazol-3-yl methanamine, 1,5-dimethyl-1h-pyrazol-3-yl methanamine 2hcl
CC1=CC(=NN1C)CN
125.175
125.17
5g
96.5
97%
MFCD03422531
JGYXJOBBROGMLL-UHFFFAOYSA-N
(1,5-dimethylpyrazol-3-yl)methanamine
2776367
97%

Chemical Identifiers

423768-52-9
125.175
JGYXJOBBROGMLL-UHFFFAOYSA-N
2776367
CC1=CC(=NN1C)CN
C6H11N3
MFCD03422531
1,5-dimethyl-1h-pyrazol-3-yl methanamine, 1,5-dimethyl-1h-pyrazol-3-yl methylamine, 3-aminomethyl-1,5-dimethylpyrazole, 1-1,5-dimethyl-1h-pyrazol-3-yl methanamine, 1,5-dimethylpyrazol-3-yl methanamine, 3-aminomethyl-1,5-dimethyl-1h-pyrazole, 1,5-dimethylpyrazol-3-yl methylamine, 1h-pyrazole-3-methanamine,1,5-dimethyl, 1-1,5-dimethylpyrazol-3-yl methanamine, 1,5-dimethyl-1h-pyrazol-3-yl methanamine 2hcl
(1,5-dimethylpyrazol-3-yl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents