missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,5-Dihydroxynaphthalene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0459930

 View more versions of this product

Catalog No. AAL0459930


Description

Description

1,5-Dihydroxynaphthalene is an intermediate of synthetic mordant azo dyes. It is an intermediate used in organic synthesis, pharmaceuticals, dyestuff fields and photograph industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dihydroxynaphthalene is an intermediate of synthetic mordant azo dyes. It is an intermediate used in organic synthesis, pharmaceuticals, dyestuff fields and photograph industry.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

1,5-Dihydroxynaphthalene
∽260°C (decomposition)
C10H8O2
250 g
1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990
BOKGTLAJQHTOKE-UHFFFAOYSA-N
naphthalene-1,5-diol
6749
98%
83-56-7
252°C (486°F)
MFCD00003980
2044951
Soluble in water.
C1=CC2=C(C=CC=C2O)C(=C1)O
160.172
160.17

Chemical Identifiers

83-56-7
160.172
BOKGTLAJQHTOKE-UHFFFAOYSA-N
6749
C1=CC2=C(C=CC=C2O)C(=C1)O
C10H8O2
MFCD00003980
1,5-dihydroxynaphthalene, 1,5-naphthalenediol, 1,5-dihydroxy naphthalene, oxidation base, durafur developer e, 1,5-dihydroxynapthalene, unii-p25hc23vh6, 1,5-dihydroxynaphtalene, 1.5-naphthalenediol, pubchem22990
naphthalene-1,5-diol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c

H302-H317-H319

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 201-487-4

RTECSNumber : QJ4740000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only