missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Dihydroxyanthraquinone, tech. 90%

$32.84 - $248.28

Chemical Identifiers

CAS 117-12-4
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.214
MDL Number MFCD00001210
InChI Key JPICKYUTICNNNJ-UHFFFAOYSA-N
Synonym anthrarufin, 1,5-dihydroxyanthraquinone, 1,5-dihydroxyanthrachinon, 9,10-anthracenedione, 1,5-dihydroxy, 1,5-dihydroxy-9,10-anthraquinone, 1,5-dihydroxy-9,10-anthracenedione, anthraquinone, 1,5-dihydroxy, az-u, unii-kpb60w5s3m, ccris 3150
PubChem CID 8328
ChEBI CHEBI:37501
IUPAC Name 1,5-dihydroxyanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)O)C(=O)C3=C(C2=O)C(=CC=C3)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0084809 View Documents Alfa Aesar™
L0084809
10g Each for $32.84
 
AAL0084822 View Documents Alfa Aesar™
L0084822
100g Each for $62.00
 
AAL0084836 View Documents Alfa Aesar™
L0084836
500g Each for $248.28
 
Specifications

Chemical Identifiers

117-12-4
240.214
JPICKYUTICNNNJ-UHFFFAOYSA-N
8328
1,5-dihydroxyanthracene-9,10-dione
C14H8O4
MFCD00001210
anthrarufin, 1,5-dihydroxyanthraquinone, 1,5-dihydroxyanthrachinon, 9,10-anthracenedione, 1,5-dihydroxy, 1,5-dihydroxy-9,10-anthraquinone, 1,5-dihydroxy-9,10-anthracenedione, anthraquinone, 1,5-dihydroxy, az-u, unii-kpb60w5s3m, ccris 3150
CHEBI:37501
C1=CC2=C(C(=C1)O)C(=O)C3=C(C2=O)C(=CC=C3)O

Specifications

~280°C
117-12-4
MFCD00001210
14,689
JPICKYUTICNNNJ-UHFFFAOYSA-N
1,5-dihydroxyanthracene-9,10-dione
8328
240.21
Technical
10g
C14H8O4
1881718
anthrarufin, 1,5-dihydroxyanthraquinone, 1,5-dihydroxyanthrachinon, 9,10-anthracenedione, 1,5-dihydroxy, 1,5-dihydroxy-9,10-anthraquinone, 1,5-dihydroxy-9,10-anthracenedione, anthraquinone, 1,5-dihydroxy, az-u, unii-kpb60w5s3m, ccris 3150
C1=CC2=C(C(=C1)O)C(=O)C3=C(C2=O)C(=CC=C3)O
240.214
CHEBI:37501
90%
1,5-Dihydroxyanthraquinone
Safety and Handling

Safety and Handling

RTECSNumber : CB6630000

TSCA : TSCA