Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Difluoro-2,4-dinitrobenzene, 97%

Chemical Identifiers

Chemical Name or Material 1,5-Difluoro-2,4-dinitrobenzene
CAS 327-92-4
Molecular Formula C6H2F2N2O4
Formula Weight 204.09
MDL Number MFCD00007052
UN Number UN2811
Beilstein 1883116
Synonym FFDNB
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2161506

 
alfa aesar™
B2161506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2161514

 
alfa aesar™
B2161514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.09
Chemical Name or Material 1,5-Difluoro-2,4-dinitrobenzene
TSCA TSCA
Assay 97%
Molecular Formula C6H2F2N2O4
MDL Number MFCD00007052
CAS 327-92-4
Health Hazard 2 GHS H Statement
H300-H311-H315-H319-H317
Fatal if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number CZ5663200
Melting Point 72° to 76°C
UN Number UN2811
Beilstein 1883116
Synonym FFDNB
Health Hazard 1 Danger
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H311-H315-H319-H317
Fatal if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.