Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Dichloroanthraquinone, 96%

$286.31 - $286.31

Chemical Identifiers

Chemical Name or Material 1,5-Dichloroanthraquinone
CAS 82-46-2
Molecular Formula C14H6Cl2O2
Formula Weight 277.11
MDL Number MFCD00001190
Beilstein 402592
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1953430

  SDS
alfa aesar™
A1953430
250g Each for $286.31
Description & Specifications

Specifications

Formula Weight 277.11
Chemical Name or Material 1,5-Dichloroanthraquinone
TSCA TSCA
Assay 96%
Molecular Formula C14H6Cl2O2
MDL Number MFCD00001190
CAS 82-46-2
Beilstein 402592
Melting Point 245° to 249°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
RTECS Number CB6495000
Quantity 250g

Applications
1,5-Dichloroanthraquinone undergoes a solid-state transformation at 470 K. The two modifications are very similar, except that a is greater than b above the transition temperature and the inverse is at room temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.