Learn More
1,5-Diazabicyclo[4.3.0]non-5-ene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1343722
Description
1,5-Diazabicyclo[4.3.0]non-5-ene is employed for dehydrohalogenation reactions and base-catalyzed rearrangements in organic synthesis. It is an amidine base used in organic synthesis. Further, it is used as a resin curing agent and polyurethane catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,5-Diazabicyclo[4.3.0]non-5-ene is employed for dehydrohalogenation reactions and base-catalyzed rearrangements in organic synthesis. It is an amidine base used in organic synthesis. Further, it is used as a resin curing agent and polyurethane catalyst.
Solubility
Miscible with water.
Notes
Air, light and moisture sensitive. Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
Specifications
1,5-Diazabicyclo[4.3.0]non-5-ene | |
1.042 | |
94°C (201°F) | |
1.52 | |
100 g | |
2417 | |
1,5-diazabicyclo 4.3.0 non-5-ene, 2,3,4,6,7,8-hexahydropyrrolo 1,2-a pyrimidine, unii-978m4ol12q, 2h,3h,4h,6h,7h,8h-pyrrolo 1,2-a pyrimidine, pyrrolo 1,2-a pyrimidine, 2,3,4,6,7,8-hexahydro, 1,5-diazabicyclo 4,3,0 non-5-ene, 1,5-diazabicyclo 4.3.0-5-nonene, 1,5-diazabicyclo-4.3.0 non-5-ene | |
SGUVLZREKBPKCE-UHFFFAOYSA-N | |
2,3,4,6,7,8-hexahydropyrrolo[1,2-a]pyrimidine | |
76349 | |
98% |
3001-72-7 | |
95°C to 98°C (7 mmHg) | |
C7H12N2 | |
MFCD00005554 | |
UN3267 | |
Air sensitive; Hygroscopic | |
Miscible with water. | |
C1CN2CCCN=C2C1 | |
124.19 | |
124.19 |
Chemical Identifiers
3001-72-7 | |
124.19 | |
SGUVLZREKBPKCE-UHFFFAOYSA-N | |
76349 | |
C1CN2CCCN=C2C1 |
C7H12N2 | |
MFCD00005554 | |
1,5-diazabicyclo 4.3.0 non-5-ene, 2,3,4,6,7,8-hexahydropyrrolo 1,2-a pyrimidine, unii-978m4ol12q, 2h,3h,4h,6h,7h,8h-pyrrolo 1,2-a pyrimidine, pyrrolo 1,2-a pyrimidine, 2,3,4,6,7,8-hexahydro, 1,5-diazabicyclo 4,3,0 non-5-ene, 1,5-diazabicyclo 4.3.0-5-nonene, 1,5-diazabicyclo-4.3.0 non-5-ene | |
2,3,4,6,7,8-hexahydropyrrolo[1,2-a]pyrimidine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 221-087-3
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only