Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Diaminonaphthalene, Ultrapure MALDI Matrix


For use as a matrix in MALDI mass spectrometry analysis

Quantity 5 x 10mg

Manufacturer: alfa aesar™  J67687EXK

 View more versions of this product

Catalog No. AAJ67687EXK

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 158.2
Boiling Point 200° to 210°C (5mmHg)
Melting Point 189° to 192°C
Chemical Name or Material 1,5-Diaminonaphthalene
MDL Number MFCD00004029
CAS 2243-62-1
Synonym DAN
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number QJ3400000
Quantity 5 x 10mg
Name Note Ultrapure MALDI Matrix
UN Number UN3077
Beilstein 907947
Color Gray to dark brown
Physical Form Powder
Health Hazard 1 Warning

Specifications

Chemical Name or Material 1,5-Diaminonaphthalene
CAS 2243-62-1
Name Note Ultrapure MALDI Matrix
Formula Weight 158.2
MDL Number MFCD00004029
UN Number UN3077
Beilstein 907947
Synonym DAN
Accessories

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.