Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Diaminonaphthalene, Ultrapure MALDI Matrix

For use as a matrix in MALDI mass spectrometry analysis

Chemical Identifiers

Chemical Name or Material 1,5-Diaminonaphthalene
CAS 2243-62-1
Name Note Ultrapure MALDI Matrix
Formula Weight 158.2
MDL Number MFCD00004029
UN Number UN3077
Beilstein 907947
Synonym DAN
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67687EXK

 
alfa aesar™
J67687EXK
5 x 10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.2
Boiling Point 200° to 210°C (5mmHg)
Melting Point 189° to 192°C
Chemical Name or Material 1,5-Diaminonaphthalene
MDL Number MFCD00004029
CAS 2243-62-1
Synonym DAN
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number QJ3400000
Quantity 5 x 10mg
Name Note Ultrapure MALDI Matrix
UN Number UN3077
Beilstein 907947
Color Gray to dark brown
Physical Form Powder
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.