Please login to your online account to display your discounted pricing

1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione, Tech., Maybridge

Chemical Identifiers

Chemical Name or Material 1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione
Assay Tech.
Molecular Formula C14H10N2O4
Formula Weight 270.24
MDL Number MFCD00013722
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01090EE

 
fisher scientific company
SB01090EE
Amber Glass Bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Formula Weight 270.24
Quantity 50g
Assay Tech.
Packaging Amber Glass Bottle
Molecular Formula C14H10N2O4
CAS 145-49-3
Chemical Name or Material 1,5-Diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione
MDL Number MFCD00013722
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.