Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Diacetoxypentane, 98%

$36.16 - $36.16

Chemical Identifiers

Chemical Name or Material 1,5-Diacetoxypentane
CAS 6963-44-6
Molecular Formula CH3CO2(CH2)5O2CCH3
Formula Weight 188.24
MDL Number MFCD00014985
Synonym 1,5-Pentanediol diacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2053709

  SDS
alfa aesar™
B2053709
10g Each for $36.16
Description & Specifications

Specifications

Formula Weight 188.24
Chemical Name or Material 1,5-Diacetoxypentane
TSCA TSCA
Assay 98%
Molecular Formula CH3CO2(CH2)5O2CCH3
MDL Number MFCD00014985
CAS 6963-44-6
Synonym 1,5-Pentanediol diacetate
Boiling Point 243° to 245°C
Flash Point >110°C (230°F)
Quantity 10g