Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate

Hydrogenation

$180.09 - $547.99

Chemical Identifiers

Chemical Name or Material 1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
CAS 38465-86-0
Formula Weight 845.79
MDL Number MFCD00064800
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1880902

 
alfa aesar™
1880902
0.5g Each for $180.09

AA1880904

 
alfa aesar™
1880904
2g Each for $547.99
Description & Specifications

Specifications

Formula Weight 845.79
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
Physical Form Crystalline
MDL Number MFCD00064800
CAS 38465-86-0
Solubility Information Soluble in acetone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 224°C (dec.)
Quantity 0.5g
Sensitivity Air & Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.