Please login to your online account to display your discounted pricing

Alfa Aesar™ (1,5-Cyclooctadiene)rhodium(I) 2,4-pentanedionate

$221.50 - $574.08

Chemical Identifiers

Chemical Name or Material (1,5-Cyclooctadiene)rhodium(I) 2,4-pentanedionate
CAS 12245-39-5
Molecular Formula C13H19O2Rh
Formula Weight 310.19
MDL Number MFCD00075046
Synonym (Acetylacetonato)(1,5-cyclooctadiene)rhodium(I); (1,5-Cyclooctadiene)(2,4-pentandionato)rhodium(I)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2608303

  SDS
alfa aesar™
H2608303
1g Each for $574.08

AAH26083MD

  SDS
alfa aesar™
H26083MD
250mg Each for $221.50
Description & Specifications

Specifications

Formula Weight 310.19
Chemical Name or Material (1,5-Cyclooctadiene)rhodium(I) 2,4-pentanedionate
Molecular Formula C13H19O2Rh
MDL Number MFCD00075046
CAS 12245-39-5
Synonym (Acetylacetonato)(1,5-cyclooctadiene)rhodium(I); (1,5-Cyclooctadiene)(2,4-pentandionato)rhodium(I)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 136° to 140°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number VI9292000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.