Please login to your online account to display your discounted pricing

Alfa Aesar™ (1,5-Cyclooctadiene)iridium(I) 2,4-pentanedionate

Chemical Identifiers

Chemical Name or Material (1,5-Cyclooctadiene)iridium(I) 2,4-pentanedionate
CAS 12154-84-6
Formula Weight 399.51
MDL Number MFCD08273779
Synonym Acetylacetonato(1,5-cyclooctadiene)iridium(I); Ir(acac)(cod)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4658103

  SDS
alfa aesar™
4658103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (1,5-Cyclooctadiene)iridium(I) 2,4-pentanedionate
Color Yellow
Melting Point 150° to 155°C (dec.)
Quantity 1g
CAS 12154-84-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number NO3615000
Physical Form Crystalline Solid
Formula Weight 399.51
MDL Number MFCD08273779
Synonym Acetylacetonato(1,5-cyclooctadiene)iridium(I); Ir(acac)(cod)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.