missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) Hexafluorophosphate, 97%, Thermo Scientific Chemicals

(1, 5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate, 97%, C34H38IrP2.F6P, CAS Number-38465-86-0

$208.37 - $208.37

Chemical Identifiers

CAS 38465-86-0
Molecular Formula C34H38F6IrP3
Molecular Weight (g/mol) 845.81
MDL Number MFCD00064800
InChI Key LXKHQEQLBSJJCO-JXNOXZOESA-N
Synonym 1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate, 1,5-cyclooctadiene bis methyldiphenylphosphine ir, 1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate, 1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate, iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate
PubChem CID 5702663
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate
SMILES [Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC377942500
SDS View Documents Promotion Details
Thermo Scientific Chemicals
377942500
250 mg Glass bottle
Each for $208.37
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38465-86-0
845.81
LXKHQEQLBSJJCO-JXNOXZOESA-N
5702663
[Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1
C34H38F6IrP3
MFCD00064800
1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate, 1,5-cyclooctadiene bis methyldiphenylphosphine ir, 1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate, 1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate, iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate
(1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate

Specifications

38465-86-0
100.0
Red
96% min
C34H38F6IrP3
250 mg
LXKHQEQLBSJJCO-JXNOXZOESA-N
(1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate
5702663
97%
(1, 5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
96.0
224.0°C
Authentic
Glass bottle
MFCD00064800
1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate, 1,5-cyclooctadiene bis methyldiphenylphosphine ir, 1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate, 1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate, iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate
[Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1
845.81
845.81
Crystalline Powder
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth.
Do

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only