Learn More
(1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) Hexafluorophosphate, 97%, Thermo Scientific Chemicals
(1, 5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate, 97%, C34H38IrP2.F6P, CAS Number-38465-86-0
Supplier: Thermo Scientific Chemicals 377942500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(1, 5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate | |
96.0 | |
224.0°C | |
Authentic | |
Glass bottle | |
MFCD00064800 | |
1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate, 1,5-cyclooctadiene bis methyldiphenylphosphine ir, 1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate, 1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate, iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate | |
[Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1 | |
845.81 | |
845.81 | |
Crystalline Powder |
38465-86-0 | |
100.0 | |
Red | |
96% min | |
C34H38F6IrP3 | |
250 mg | |
LXKHQEQLBSJJCO-JXNOXZOESA-N | |
(1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate | |
5702663 | |
97% |
Chemical Identifiers
38465-86-0 | |
845.81 | |
LXKHQEQLBSJJCO-JXNOXZOESA-N | |
5702663 | |
[Ir+].F[P-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1 |
C34H38F6IrP3 | |
MFCD00064800 | |
1,5-cyclooctadiene bis methyldiphenylphosphine iridium i hexafluorophosphate, 1,5-cyclooctadiene bis methyldiphenylphosphine ir, 1,5-cyclooctadienebis methyldiphenylphosphine iridium i hexafluorophosphate, 1z,5z-cycloocta-1,5-diene; iridium; methyl diphenyl phosphane; hexafluorophosphate, iridium 1+ 1,5-cyclooctadiene, z,z-bis methyldiphenylphosphane hexafluorophosphate | |
(1Z,5Z)-cycloocta-1,5-diene;iridium;methyl(diphenyl)phosphane;hexafluorophosphate |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth.
Do
GHS Signal Word: Warning
RUO – Research Use Only