Please login to your online account to display your discounted pricing

1-(5-Chloro-2-hydroxyphenyl)-3-phenyl-1,3-propanedione, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(5-Chloro-2-hydroxyphenyl)-3-phenyl-1, 3-propanedione
CAS 5067-25-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H11ClO3
Linear Formula C6H5COCH2COC6H3(Cl)OH
Formula Weight 274.7
MDL Number MFCD00209562
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372960010

  SDS
thermo fisher scientific
AC372960010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC372960050

  SDS
thermo fisher scientific
AC372960050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Needles
Formula Weight 274.7
Quantity 1g
Color Yellow
Melting Point 107° to 112°C
CAS Max % 100.0
Infrared Spectrum Authentic
MDL Number MFCD00209562
CAS Min % 97.5
Packaging Glass bottle
Linear Formula C6H5COCH2COC6H3(Cl)OH
Chemical Name or Material 1-(5-Chloro-2-hydroxyphenyl)-3-phenyl-1, 3-propanedione
Molecular Formula C15H11ClO3
CAS 5067-25-4
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.