Please login to your online account to display your discounted pricing

1-(5-Bromo-2-hydroxyphenyl)-3-phenyl-1,3-propanedione, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC372970010

 View more versions of this product

Catalog No. AC372970010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 319.15
Quantity 1g
Color Yellow
CAS Max % 100.0
Chemical Name or Material 1-(5-Bromo-2-hydroxyphenyl)-3-phenyl-1, 3-propanedione
Assay Percent Range 98%
Molecular Formula C15H11BrO3
Linear Formula C6H5COCH2COC6H3(Br)OH
MDL Number MFCD00209552
CAS 5067-24-3
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1-(5-Bromo-2-hydroxyphenyl)-3-phenyl-1, 3-propanedione
CAS 5067-24-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H11BrO3
Linear Formula C6H5COCH2COC6H3(Br)OH
Formula Weight 319.15
MDL Number MFCD00209552
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents