Please login to your online account to display your discounted pricing

1-(5-Bromo-2,3-dihydro-1H-indol-1-yl)ethan-1-one, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  KM08258DA

 View more versions of this product

Catalog No. KM08258DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 240.1
Quantity 1g
Assay 97%
CAS Min % 97.0
Molecular Formula C10H10BrNO
CAS 22190-38-1
MDL Number MFCD00056017
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-(5-Bromo-2, 3-dihydro-1H-indol-1-yl)ethan-1-one
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 1-(5-Bromo-2, 3-dihydro-1H-indol-1-yl)ethan-1-one
CAS 22190-38-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H10BrNO
Formula Weight 240.1
MDL Number MFCD00056017
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.