missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,5-Bis(methylamino)-3-oxapentane, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC275860050

 View more versions of this product

Catalog No. AC275860050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 5-Bis(methylamino)-3-oxapentane
Authentic
1.4370 to 1.4390
76.0°C to 78.0°C (20.0mmHg)
5g
98%
CH3NHCH2CH2OCH2CH2NHCH3
Solubility in water: soluble. Other solubilities: soluble in methanol,chloroform
CNCCOCCNC
132.21
132.21
98%
0.8720g/mL
Glass bottle
0.872
Yellow
2620-27-1
C6H16N2O
1,5-bis methylamino-3-oxapentane, 2,2'-oxybis n-methylethanamine, bmaop, ethanamine, 2,2'-oxybis n-methyl, ethanamine,2,2'-oxybis n-methyl, methyl 2-2-methylamino ethoxy ethyl amine, n-methyl-2-2-methylamino ethoxy ethanamine, n-methyl-2-methylaminoethyloxy ethylamine, n-methyl-n-2-2-methylamino ethoxy ethyl amine
VXPJBVRYAHYMNY-UHFFFAOYSA-N
N-methyl-2-[2-(methylamino)ethoxy]ethanamine
2724770
Liquid

Chemical Identifiers

2620-27-1
132.21
1,5-bis methylamino-3-oxapentane, 2,2'-oxybis n-methylethanamine, bmaop, ethanamine, 2,2'-oxybis n-methyl, ethanamine,2,2'-oxybis n-methyl, methyl 2-2-methylamino ethoxy ethyl amine, n-methyl-2-2-methylamino ethoxy ethanamine, n-methyl-2-methylaminoethyloxy ethylamine, n-methyl-n-2-2-methylamino ethoxy ethyl amine
N-methyl-2-[2-(methylamino)ethoxy]ethanamine
C6H16N2O
VXPJBVRYAHYMNY-UHFFFAOYSA-N
2724770
CNCCOCCNC
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS Documents