missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,5-Bis(diphenylphosphino)pentane, 97%, ACROS Organics™

$47.19 - $47.19

Chemical Identifiers

CAS 27721-02-4
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C29H30P2
Formula Weight 440.51
MDL Number MFCD00003052
Chemical Name or Material 1, 5-Bis(diphenylphosphino)pentane
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC320850050 SDS Thermo Fisher Scientific
AC320850050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC320850010 SDS Thermo Fisher Scientific
320850010
Glass bottle 1g Each for $47.19
Specifications Description & Specifications

Chemical Identifiers

27721-02-4
100.0
C29H30P2
MFCD00003052
≥96.0%
96.0
97%
440.51
1, 5-Bis(diphenylphosphino)pentane

Specifications

27721-02-4
100.0
Glass bottle
97%
440.51
White to light yellow
5g
≥96.0%
96.0
Authentic
Powder to Crystals or Chunks
C29H30P2
MFCD00003052
42° to 47°C
1, 5-Bis(diphenylphosphino)pentane

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.