Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,5-Bis(4-aminophenoxy)pentane, 97%

$50.78 - $705.16

Chemical Identifiers

Chemical Name or Material 1,5-Bis(4-aminophenoxy)pentane
CAS 2391-56-2
Molecular Formula C17H22N2O2
Formula Weight 286.37
MDL Number MFCD01632139
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2143403

  SDS
alfa aesar™
B2143403
1g Each for $50.78

AAB2143414

  SDS
alfa aesar™
B2143414
25g Each for $705.16
Description & Specifications

Specifications

Formula Weight 286.37
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
CAS 2391-56-2
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD01632139
Quantity 1g
Molecular Formula C17H22N2O2
Chemical Name or Material 1,5-Bis(4-aminophenoxy)pentane
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.