Please login to your online account to display your discounted pricing

1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one, 97%, Maybridge

$345.48 - $345.48

Chemical Identifiers

Chemical Name or Material 1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Assay 95%
Assay Percent Range 95%
Molecular Formula C16H22O
Formula Weight 230.35
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00212EA

 
fisher scientific company
TL00212EA
Amber glass bottle 10g Each for $345.48
Description & Specifications

Specifications

Formula Weight 230.35
Quantity 10g
Assay 95%
Chemical Name or Material 1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Packaging Amber glass bottle
Assay Percent Range 95%
CAS 17610-21-8
Molecular Formula C16H22O