Please login to your online account to display your discounted pricing

1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  TL00212EA

 View more versions of this product

Catalog No. TL00212EA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 230.35
Quantity 10g
Assay 95%
Chemical Name or Material 1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Packaging Amber glass bottle
Assay Percent Range 95%
CAS 17610-21-8
Molecular Formula C16H22O

Specifications

Chemical Name or Material 1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Assay 95%
Assay Percent Range 95%
Molecular Formula C16H22O
Formula Weight 230.35